missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ U-Chem 3200 Chemical Protection Gloves

Product Code. 17240531
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

NBR-coated chemical protection gloves feature a slim-fit design to provide high levels of dexterity. These gloves are ideal for handling oil, cooling lubricants, and chemicals.

  • NBR-coated chemical protection gloves with nylon weave supporting material
  • Slim-fit design easily stretches and naturally moves with the hand for increased dexterity
  • Additional coating on the palm ensures a secure grip
  • Resistant to grease, mineral oils, and various chemicals
  • Ideal for handling oil, cooling lubricants, and grease
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Coating Material NBR
Color Blue, Black
Size 9
EN388 Standard 3131X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLMOT
For Use With (Application) Handling Oil, Cooling Lubricants, and Chemicals
Gender Unisex
Glove Cuff Style Gauntlet
Liner Material Nylon
Series U-Chem 3200
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'