missing translation for 'onlineSavingsMsg'
Learn More

UK 5099, Tocris Bioscience™

Product Code. 18094302
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MCT inhibitor; also inhibits mitochondrial pyruvate carrier

Inhibitor of plasma membrane monocarboxylate transporters (MCTs) and the mitochondrial pyruvate carrier (MPC).

Chemical Identifiers

CAS 56396-35-1
Molecular Formula C18H11N2O2-
Molecular Weight (g/mol) 287.298
InChI Key BIZNHCWFGNKBBZ-JLHYYAGUSA-M
Synonym 2-cyano-3-1-phenyl-1h-indol-3-yl-2-propenoic acid, 2e-2-cyano-3-1-phenylindol-3-yl prop-2-enoate
PubChem CID 71772548
IUPAC Name (E)-2-cyano-3-(1-phenylindol-3-yl)prop-2-enoate
SMILES C1=CC=C(C=C1)N2C=C(C3=CC=CC=C32)C=C(C#N)C(=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 288.3
Percent Purity >99%
Chemical Name or Material UK 5099
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'