Learn More
Greiner Bio-One Ultra Low Profile Polystyrene Microplate Lids
Used for high-format low profile microplates
226.00€
Specifications
Color | Clear |
---|---|
Material | Polystyrene |
Condensation Rings | No |
No. per Case | 100 |
No. per Pack | 5 |
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic, non-cytotoxic
- Offer protection against contamination and evaporation during sample storage and cell cultivation
- Compatible with a range of different microplates
Specifications
Clear | |
No | |
5 |
Polystyrene | |
100 | |
A variety of microplates, particularly well suited for high-format, low-profile microplates |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.