Learn More
uvex Unidur 6641 Cut Protection Gloves
Shop All Uvex Products
Click to view available options
Size:
6
11
Description
- Lightweight, polyurethane-coated HPPE assembly gloves
- Coating provides mechanical abrasion resistance and level 3/B cut protection
- Enhanced grip in dry or slightly damp areas
- High level of sensitivity for tasks that require tactility
Specifications
Specifications
Coating Material | Polyurethane |
Color | White, Gray |
Hand | Ambidextrous |
Latex Free | Yes |
Material | HPPE, Elastane |
Product Type | Cut-resistant Gloves |
Size | 11 |
EN388 Standard | 4343B |
Glove Cuff Style | Knit |
Series | Unidur 6641 |
Product Content Correction
Product Title
uvex Unidur 6641 Cut Protection Gloves >