missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey Nagel UNISOL pH Indicator Solution
For pH determination in pure water, in surface waters, and in very dilute acids or bases.
Brand: Macherey Nagel 91031
This item is not returnable.
View return policy
Description
- pH determinations in weakly buffered solutions
- UNISOL indicator solutions are used for pH determination in pure water, in surface waters and in very dilute acids or bases.
- An indicator solution is added to the sample and the reaction color is compared with a color scale.
- Therefore, they allow the easy and reliable pH determination in weakly buffered solutions.
Specifications
1 to 13 | |
Color chart, cuvette | |
pH Indicator Solution |
100 mL | |
1 Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction