missing translation for 'onlineSavingsMsg'
Learn More

ZVG™ Unitex™ Wipes

These convenient cleaning towels are made from viscose nonwoven material with textile-like properties and have a high liquid absorption capacity.

Brand:  ZVG™ 10148-01

Product Code. 17823675

  • 22.85€ / Pack of 200
Estimated Shipment: 05-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Viscose fibers, produced industrially from cellulose, give the fabric its 'natural synthetic' textile character. The chemical composition of this material is similar to that of cotton. Viscose products are absorbent, flexible, and durable.

  • Can be used both dry and wet
  • Ideal for cleaning and degreasing surfaces, such as those to be painted (in conjunction with cleaning agents like silicone remover, water-based cleaners, or water)
  • Good for use with 2K paints and water-based paints
  • Tear- and solvent-resistant
  • Silicone-free
  • Lint-free
Specifications

Specifications

Wipe
38 cm
30 cm
Turquoise
Non-sterile
15 in.
11.8 in.
Polyester Cellulose
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ZVG™ Unitex™ Wipes > Unitex Wipes; Color: Turquoise; Material: Polyester Cellulose

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.