Learn More
Thermo Scientific™ Accessories for Digital Mini Rotators
Get assorted accessories for use with Digital Mini Rotators
Brand:  Thermo Scientific™ 88882113
126.63 EUR valid until 2025-06-30
Use promo code "24108" to get your promotional price.
 View more versions of this product
Product Code. 16584792
  • 126.63€ / Each
In Stock
Description
Assorted accessories for use with Digital Mini Rotators
Service Plan
Standard Thermo warranty guideline, provide repair service and free spare part within warranty
Services
2 years
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flat Platform with Rubber Mat
330 x 270 mm
Flat Platform w/ Rubber Mat
Stainless Steel, rubber
330 x 270 x 19 mm
Digital Mini Rotators, Rocker, Rotator, Mixer
Product Title
Thermo Scientific™ Accessories for Digital Mini Rotators >