Learn More
Fisherbrand™ Universal pH Indicator Paper
Product Code. p-8772035 Shop All Fisherbrand Products
Click to view available options
pH Range:
0 to 10
1 to 14
5 to 9
Unit Size:
Each
Product Code. 11703564
Edge
Provides rapid and precise readings. Fisherbrand™ Universal pH Indicator Paper include a color scale printed on the cover for convenience.
  • Filter paper soaked with one or several colored indicators of pH
  • Economic range of pH 0 to 14
  • Available in a variety of formats to meet individual needs
  • Reference chart shows color changes in steps of pH2
Specifications
pH Range 5 to 9
Format Roll
Quantity 1 Roll
Type Grade 911
Length (Metric) 5 m
Width (Metric) 7 mm