Learn More
Kinesis™ Back Pressure Regulators: Cartridges
For use with Upchurch Scientific™ Back Pressure Regulators
160.32€
Product Code | Brand | Pressure | Swept Volume | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pressure | Swept Volume | Price | Quantity & Availability | ||||
13080410
|
Kinesis™
02017-16 |
75 psi | 125 μL |
160.32€
Each |
In Stock
Log in to see stock. |
||||
13070410
|
Kinesis™
02017-14 |
40 psi | 125 μL |
160.32€
Each |
In Stock
Log in to see stock. |
||||
Description
Biocompatible/chemically resistant materials.
Specifications
Cartridge | |
Back Pressure Regulators |