Learn More
WTW™ USB Charger for IDS Wireless Modules
The WTW™ USB Charger for IDS Wireless Modules is used with all instruments with wireless measurement capability of pH, ORP, oxygen, conductivity and turbidity.
Brand:  WTW™ 108143
Product Code. 15532016
  • 138.00€ / Each
Estimated Shipment: 03-01-2025
Description
  • For recharging sensor modules used with wireless-ready portable meters
  • Can be used with or without electrode connected
  • Suitable for wall mounting
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USB Charger
IDS Wireless Modules