Learn More
Cytiva USB to RS232 Adapter for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
USB to RS232 Adapter for Ettan™ IPGphor™ 3 Isoelectric Focusing Unit
Brand: Cytiva 28-4001-45
Description
- To connect RS232 to USB port of computer
- For use with Ettan IPGphor 3 isoelectric focusing unit
Specifications
Ettan IPGphor 3 IEF Unit | |
USB to RS-232 Adapter | |
Complete packsize |