missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP53 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
Antigen | USP53 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18350253
|
Bio-Techne
NBP3-05172-20UL |
20 μL |
198.00€
20µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
18392213
|
Bio-Techne
NBP3-05172-100UL |
100 μL |
468.00€
100µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
USP53 Polyclonal specifically detects USP53 in Human samples. It is validated for Western Blot.Specifications
USP53 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
54532 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
DKFZp781E1417, inactive ubiquitin carboxyl-terminal hydrolase 53, Inactive ubiquitin-specific peptidase 53, KIAA1350, ubiquitin specific peptidase 53, ubiquitin specific protease 53, ubiquitin specific proteinase 53 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 886-1073 of human USP53 (NP_061923.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title