Learn More
Vilber Lourmat UV Protection Face Shield
Product Code. 11726778
Click to view available options
Product Type:
UV-protection faceshield
Rich, robust UV (ultraviolet) protection face shield that fully protects the face.
  • UV radiation
  • Spectral curves of tubes
  • 254nm = UV-C
  • High bactericidal power transmitted only through quartz
  • 312nm = UV-B
  • 365nm = UV-A
  • BLB version called Black Light or Wood Light, black tube without visible light
  • User protection Imperative regardless of the wavelength emitted
  • UV protection face shield that fully protects the face
  • For work on light table
Specifications
Description UV Protection Face Shield
Product Type UV-protection faceshield
Model MP-80 UV protection mask