Learn More
Syngene™ UV Safety Shield
Safety shield designed to offer UV protection.
189.11€ - 206.74€
Specifications
Voltage | 110/230 V |
---|---|
Product Line | Syngene |
For Use With (Application) | UV Protection |
Product Type | Safety Shield |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16534351
![]() |
Syngene™
UV-SHIELD-SMALL |
Small |
189.11€
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||||
16524351
![]() |
Syngene™
UV-SHIELD-LARGE |
Large |
206.74€
Each |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
Description
- The wearing of safety equipment according to laboratory regulations is recommended
- Small Shield Dimensions: 14 W x 19 in. H
- Large Shield Dimensions: 14 W x 24 in. H
Specifications
110/230 V | |
Syngene | |
UV Protection | |
Safety Shield |
Product Title
Syngene™ UV Safety Shield