Learn More
BD Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube
Used to obtain a whole blood or EDTA plasma sample. Vacutainer™ Hemogard™ Closure Plastic K2-Edta Tube contains K2EDTA which is a liquid solution that is spray coated onto the interior surface of the BD Vacutainer Plus Tubes.
Brand: BD 367873
Description
Lavender Hemogard™ closurePaper label
Specifications
K2 EDTA Tube | |
Plastic | |
6 mL | |
100 mm | |
13 mm |
K2 EDTA | |
Hemogard | |
Lavender | |
Block |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H332-Harmful if inhaled.
Precautionary Statement
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.