Learn More
BD Vacutainer™ Syringe Critical Care Blood Collection
Brand:  BD 364390
Product Code. 12907766
  • 298.00€ / Pack of 100
Estimated Shipment: 25-11-2024
Description
with safety needle BD preset, luer-lok with BD Hemogard™ tip cap 22G BD Eclipse volume 3mL BD Vacutainer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
22 G
Eclipse™ safety needle, Calcium-balanced lithium heparin spray dried on the sides of the syringe
3 mL
Measure arterial blood gas, ionized calcium, ionized magnesium, as well as perform critical care tests
1 in.
Luer Lock