Learn More
WTW™ Variable Wave-length Thermoreactor with 4 Temperature Settings
For the disintegration of COD, total nitrogen and total phosphorus
Brand: WTW™ 1P22-
1650.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Eight fixed standard programs for simple operation
- Eight user-defined programs of 25-170°C with high temperature digestion
- AQA/IQC with external temperature sensor
Specifications
Thermoreactor | |
230/115 V | |
315 mm | |
Routine and specialized tasks in wastewater and in laboratories | |
2 x 12 Reaction Cuvettes |
50/60 Hz | |
560 W | |
115/230 V ±15%, 50/60 Hz, 560 W | |
185 mm | |
256 mm |
Product Content Correction
Product Title
WTW™ Variable Wave-length Thermoreactor with 4 Temperature Settings > Capacity: 2 x 12 Reaction cuvettes