Learn More
Thermo Scientific Chemicals Vascular Endothelial Growth Factor-165, mouse
148.74€ - 176.00€
Specifications
Molecular Weight (g/mol) | 38.8 kd |
---|---|
Packaging | Plastic bottle |
Form | Lyophilized |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
38.8 kd | |
Plastic bottle | |
Lyophilized |
RUO – Research Use Only