missing translation for 'onlineSavingsMsg'
Learn More

VDAC2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Bio-Techne NBP2-97576F

Product Code. 18101290

  • 414.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

VDAC2 Polyclonal specifically detects VDAC2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VDAC2
Polyclonal
PBS
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
FITC
FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2
Produced in rabbits immunized with E. coli-derived Human VDAC2 fragment.
0.1 mL
Lipid and Metabolism
7417
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VDAC2 Antibody, FITC, Novus Biologicals™ > 0.1 mL; FITC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.