missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Veliparib, Thermo Scientific™
Click to view available options
Quantity:
100 mg
Specifications
Specifications
CAS | 912444-00-9 |
Quantity | 100 mg |
Molecular Formula | C13 H16 N4O |
Formula Weight | 244.30 |
MDL Number | MFCD16661059 |
Physical Form | Powder |
Color | White to Yellow |
Purity | ≥98.5% |
InChI Key | JNAHVYVRKWKWKQ-UHFFFAOYNA-N |
SMILES | CC1(CCCN1)C1=NC2=C(C=CC=C2N1)C(N)=O |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction