Learn More
Cole-Parmer™ Venturi Miniature Vacuum Pump
The remarkably compact size of these Cole-Parmer Venturi Miniature Vacuum Pump enables you to design vacuum systems with smaller, less expensive components and reduced maintenance costs. With no moving parts or electrical hazards, these air-powered pumps are also intrinsically safe.
Brand:  Cole-Parmer™ 78165-40
Product Code. 11770079
  • 129.00€ / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • No moving parts ensures long life with low maintenance.
  • Venturi pumps are air-powered, require no electrical power source or lubricant.
  • Place these compact vacuum pumps close to application for fast response and minimal vacuum loss.
Specifications
Specifications
100 Mbar
Aluminum
8.5 L/min.
Vacuum System