Learn More
3M™ Versaflo™ Breathing Tubes for Headtops and Air Delivery Units
Click to view available options
Size:
Medium/Large
Small/Medium
Unit Size:
Each
Description
- Long, lightweight breathing tube that is easy to wipe down
- Quick release swivel enables one-handed connection and release
- Disposable breathing tube cover is available to help cleaning and decontamination operations
- 3M Catalog# BT-20S and BT-20L
Specifications
Specifications
Product Type | Breathing Tube |
Size | Small/Medium |
For Use With (Application) | Assembly, facility maintenance, grinding, machine operation, painting, sanding |
For Use With (Equipment) | Versaflo™ PAPR Systems |
Model | BT-20S |
Length (English) | 28 in. |
Length (Metric) | 71.1 cm |
Product Content Correction
Product Title
3M™ Versaflo™ Breathing Tubes for Headtops and Air Delivery Units >