Learn More
3M™ Versaflo™ Comfort Pad for S-950 Premium Head Suspension
Click to view available options
Size:
Small
Standard
Unit Size:
Pack of 5
Description
- For use with S-950 Premium Head Suspension
- Wrapped around headband to create more comfortable fit and to absorb sweat
- 3M Catalog# S-953 , 3M Catalog# S-954
Specifications
Specifications
Color | Gray |
Disposable | Reusable |
Product Type | Head Suspension |
Size | Standard |
For Use With (Application) | Cleaning, Electrical, Facility Maintenance, Welding |
For Use With (Equipment) | S-950 Premium Head Suspension |
Model | S-953 |
Product Content Correction
Product Title
3M™ Versaflo™ Comfort Pad for S-950 Premium Head Suspension >