Learn More
Saint Gobain Life Sciences Versilon™ SPX-50 Tubing
Versilon™ SPX-50 Tubing is a flexible silicone tubing.
Brand: Saint Gobain Life Sciences ABX00022
This item is not returnable.
View return policy
Description
- Versilon™ SPX-50 tubing from Saint-Gobain is peroxide-cured and designed for use in applications where flexibility resiliency and durability are required.
- Produced from a proprietary combination of silicon elastomers, this flexible tubing optimizes critical physical properties such as tensile strength elongation and compression set, resulting in a more physically durable product.
- The smooth inner surface of the tubing reduces the risk of particulate entrapment and microscopic buildup during fluid transfer.
Specifications
Tubing | |
Versilic | |
Flexible |