missing translation for 'onlineSavingsMsg'
Learn More

Proteintech Viability/Cytotoxicity Assay Kit for Animal Live & Dead Cells (Calcein AM, EthD-1 Method)

Viability/Cytotoxicity Assay Kit for Animal Live Dead Cells (Calcein AM, EthD-I method) provides dual fluorescence staining for the detection of living and dead cells.

105.00€ - 199.00€

Specifications

For Use With (Application) Flow Cytometry
Emission 617 nm, 517 nm
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17571176
View Documents
Proteintech
PF00008-150T
150 Tests
105.00€
150 tests
In Stock
to see stock.
Add to basket
 
17581176
View Documents
Proteintech
PF00008-300T
300 Tests
199.00€
300 tests
In Stock
to see stock.
Add to basket
 
Description

Description

  • The two probes in the kit effect cell viability by measuring intracellular esterase activity and plasma membrane integrity.
  • This kit can be used for fluorescence microscopes, flow cytometers, microplate readers and other fluorescence detection systems.
  • This kit can be used for most eukaryotic mammalian samples, but not to fungi and yeast.
Specifications

Specifications

Flow Cytometry
617 nm, 517 nm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.