Learn More
Thermo Scientific™ Vial, Cuvette and Test Tube Holders for Orion™ AQ7100 and AQ8100 Spectrophotometers
Quickly adapt your Orion™ AquaMate 7100 & 8100 Spectrophotometer to work with multiple sample vessels
25.93€ - 37.49€
Specifications
For Use With (Equipment) | Orion AquaMate 7100 & 8100 Spectrophotometers |
---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
16459835
|
Thermo Scientific™
AQX1LWLVH |
Long path rectangular cell holder assembled in a removable compartment liner. | Orion AquaMate 7100 & 8100 Spectrophotometers |
37.49€
Each |
In Stock
Log in to see stock. |
|||||
16469835
|
Thermo Scientific™
AQX1SQVH |
10 mm square single-cell holder | Orion AquaMate 7100 & 8100 Spectrophotometers |
25.93€
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
|||||
Description
Easy to change & clean Orion™ AquaMate 7100 & 8100 Spectrophotometer sample holders are designed for maximum flexibility.
Easily adapt your Orion™ AquaMate 7100 & 8100 Spectrophotometer to work with multiple sample vessels
Magnetic retention makes these holders quick and easy to clean and change to adapt to your analytical needs.
- AQX1RNDVH: Vial / Test Tube Holder for vials and test tube from 12 to 25 mm OD for Orion™ AquaMate 7100 & 8100 Spectrophotometers
- AQX1SQVH: Standard 10 mm square single-cell holder for Orion™ AquaMate 7100 & 8100 Spectrophotometers
- AQX1LWLVH: Long path rectangular cell holder for 20-100 mm pathlength cuvettes for Orion™ AquaMate 7100 & 8100 Spectrophotometers
Specifications
Orion AquaMate 7100 & 8100 Spectrophotometers |