Learn More
BINDER™ Viewing Window and Interior Lighting
This Viewing Window and Interior Lighting is designed for use with assorted BINDER™ Drying and Heating Chambers. Insulated viewing window and interior lighting allows observation without interfering with internal conditions.
1055.00€ - 2515.00€
Specifications
Type | Viewing Window with Interior Lighting |
---|---|
Insulation | Triple Insulating Glass |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
16484049
|
BINDER™
8012-1058 |
BINDER Chambers ED 56 Drying and Heating Chambers with Gravity Convection, 230 V |
1070.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
16414059
|
BINDER™
8012-1064 |
BINDER Chambers ED 260 Drying and Heating Chambers with Natural Convection, 230 V |
1420.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
16444459
|
BINDER™
8012-1830 |
BINDER Chambers ED 720 Drying and Heating Chambers with Natural Convection, 400 V |
2140.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
16494139
|
BINDER™
8012-1259 |
BINDER Chambers FD 23 Drying and Heating Chambers with Forced Convection, 230 V |
1055.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
16474389
|
BINDER™
8012-1700 |
BINDER Chambers FD 56 Drying and Heating Chambers with Forced Convection, 230 V |
1070.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
16494389
|
BINDER™
8012-1708 |
BINDER Chambers FD 260 Drying and Heating Chambers with Forced Convection, 230 V |
1420.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
16454459
|
BINDER™
8012-1831 |
BINDER Chambers FD 720 Drying and Heating Chambers with Forced Convection, 400 V |
2140.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
16484389
|
BINDER™
8012-1701 |
BINDER Chambers FED 56 Drying and Heating Chambers with Forced Convection and Enhanced Timer Functions, 230 V |
1070.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
16404399
|
BINDER™
8012-1709 |
BINDER Chambers FED 260 Drying and Heating Chambers with Forced Convection and Enhanced Timer Functions, 230 V |
1420.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
16454129
|
BINDER™
8012-1239 |
BINDER Chambers FED 400 Drying and Heating Chambers with Forced Convection and Enhanced Timer Functions, 400 V |
2515.00€
Each |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
Description
- Viewing window with triple insulating glass in door(s)
- 15 or 30 W interior lighting, depending on the model
Specifications
Viewing Window with Interior Lighting | |
Triple Insulating Glass |
Product Title
BINDER™ Viewing Window and Interior Lighting