Learn More
Greiner Bio-One VIEWseal Adhesive Microplate Sealer
For protein crystallography. Greiner Bio-One VIEWseal Adhesive Microplate Sealer is special highly transparent sealer with low autofluorescence for precise optical measurements.
Brand: Greiner Bio-One 676070
This item is not returnable.
View return policy
Description
- Coated with silicone adhesive
- Non-Pyrogenic
- Highly transparent film for optical measurements
- Sticks only when film surface is pressed on
- Film does not adhere to gloves
- Temperature-resistant from -80°C to
Specifications
Transparent | |
100 | |
Silicone |
Non-sterile | |
Sealer | |
Microplate |
For research use only. Not for use in diagnostic procedures, unless otherwise indicated.