Learn More
Brady™ Vinyl Cloth Labels for M611, BMP61 and BMP71
Once applied, this repositionable vinyl cloth label is there to stay...unless you want to change it.
Brand:  Brady™ 062659
 View more versions of this product
Product Code. 12343478
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
  • Adhesive and cloth backing offers excellent holding power, while allowing for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Specifications
Specifications
Not Autoclavable
White
Vinyl Cloth (B-498)
Non-sterile
BMP™61, BMP™71, BradyPrinter M611, TLS 2200™ and TLS-PC LINKTM Thermal Labeling Systems
750 Labels
-40°C to +80°C
9.5 mm
6.4 mm
19.1 mm
Repositionable Rubber
Semi-Gloss
Thermal Transfer
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, Wire and cable Labeling
1
Rectangle
0.185 mm
16 AWG to 10 AWG
6.4 mm
19.1 mm