missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral-Plex™ Complexation Buffer, prototype
Gibco Viral-Plex Complexation Buffer, prototype, is a chemically defined, protein-free, phenol red-free buffer for complexing plasmid DNA with transfection reagent for standard transfection applications.
Brand: Invitrogen A49846DG
This item is not returnable.
View return policy
Description
- Chemically defined, protein-free, phenol red-free buffer
- For complexing plasmid DNA with transfection reagent for standard transfection applications
Specifications
HEK293 | |
100mL bottle, store at 2°C to 8°C, protected from light. Do not freeze. | |
Mammalian | |
Lipid-based Transfection | |
AAV-MAX | |
100 mL |
animal origin free, protein free, phenol red free, chemically defined | |
Suspension | |
Plasmid DNA | |
AAV Production | |
No | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures. This product is a prototype and performance characteristics of this product have not been established.