Learn More
Thermo Scientific™ VISIONlite™ Spectrophotometer Software
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Brand:  Thermo Scientific™ 869-191700
554.21 EUR valid until 2025-04-18
Use promo code "24095" to get your promotional price.
 View more versions of this product
Product Code. 17778418
  • 3210.00€ / Each
Edge
Explore more special offers
Description
Show More
Specifications
Specifications
Stand-alone software package for Evolution and GENESYS Spectrophotometers with an extensive list of predefined procedures for wine and juice analysis.
Thermo Scientific GENESYS 40, 50, 140, 150, 180, BioMate 160, Evolution 201, 220, 260 Bio, 350 Spectrophotometers
Interface cable 840-312200 for Thermo Scientific GENESYS 40-180 and BioMate 160 Spectrophotometers.
VISIONlite Wine Analysis Software
DVD with software installer and user manual (in English and German). Display languages: English, German, Spanish, Portuguese, French, Italian, Russian and Korean.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use code 24095 to receive this promotion.
    Promo Code: 24095
    Expires2025-04-15
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 17/03/2025 and 15/04/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.
    Terms & Conditions Learn More
Product Content Correction
Product Title
Thermo Scientific™ VISIONlite™ Spectrophotometer Software >