Learn More
Sartorius Vivaflow Tubing for Vivaflow Feed Reservoir
Vivaflow Tubing used with the Vivaflow feed reservoir, enables continuous diafiltration that is easy to set up, offers significant time savings, reduces reagent and water consumption, and avoids sample dilution.
Brand: Sartorius VF-ATD0001-1
20.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Supports continuous diafiltration with Vivaflow cassettes
- Securely connects to the Vivaflow feed reservoir
- Works with Sartobind Lab for chromatography with a pump
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Transparent | |
0.12 in. | |
PVC, Polycarbonate | |
6.3 mm | |
1 m | |
Cassette | |
PES | |
0.5 to 1 L | |
Transparent | |
TFF cassette |
Not Autoclavable | |
3.1 mm | |
0.24 in. | |
39.4 in. | |
Ultrafiltration | |
Peristaltic Pump | |
10 kDa | |
Non-sterile | |
Tubing | |
1 L |