Learn More
KARTELL Volumetric Flask
Volumetric Flask, made of transparent fluorinated resin, has high chemical and temperature resistance.
Brand: KARTELL 0169600
This item is not returnable.
View return policy
Description
- Volumetric flask made of transparent fluorinated resin with high chemical and temperature resistance (-250°C to +270°C).
- Supplied with standard DIN stoppers in PFA.
- Individually calibrated ringmark according to DIN EN ISO 1042, class “A”, Standards.
- Laser engraved lot number.
Specifications
50 mL | |
Not Autoclavable | |
No | |
Non-sterile | |
Non-treated |
PFA | |
Reusable | |
Narrow | |
Flat |