Learn More
Tocris Bioscience™ w-Agatoxin TK
CaV2.1 blocker
Brand:  Tocris Bioscience™ 2802/10U
Product Code. 18064402
  • 297.00€ / 10µg
Estimated Shipment: 14-03-2025
Description
ω-Agatoxin TK is a selective blocker of CaV2.1 P/Q-type calcium channels.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
>99%
5273.02
10 μg
w-Agatoxin TK
ω-Agatoxin TK
Glu-Asp-Asn-Cys-Ile-Ala-Glu-Asp-Tyr-Gly-Lys-Cys-Thr-Trp-Gly-Gly-Thr-Lys-Cys-Cys-Arg-Gly-Arg-Pro-Cys-Arg-Cys-Ser-Met-Ile-Gly-Thr-Asn-Cys-Glu-Cys-Thr-Pro-Arg-Leu-Ile-Met-Glu-Gly-Leu-Ser-Phe-Ala (Disulfide bridge: C4-C20,C12-C25,C19-C36,C27-34)
Documents
One moment while we fetch your results.
Product Title
Tocris Bioscience™ w-Agatoxin TK > 10μg