Learn More
Tocris Bioscience™ w-Agatoxin TK
CaV2.1 blocker
Brand: Tocris Bioscience™ 2802/10U
This item is not returnable.
View return policy
Description
ω-Agatoxin TK is a selective blocker of CaV2.1 P/Q-type calcium channels.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
>99% | |
5273.02 | |
10 μg |
w-Agatoxin TK | |
ω-Agatoxin TK | |
Glu-Asp-Asn-Cys-Ile-Ala-Glu-Asp-Tyr-Gly-Lys-Cys-Thr-Trp-Gly-Gly-Thr-Lys-Cys-Cys-Arg-Gly-Arg-Pro-Cys-Arg-Cys-Ser-Met-Ile-Gly-Thr-Asn-Cys-Glu-Cys-Thr-Pro-Arg-Leu-Ile-Met-Glu-Gly-Leu-Ser-Phe-Ala (Disulfide bridge: C4-C20,C12-C25,C19-C36,C27-34) |
Product Content Correction
Product Title
Tocris Bioscience™ w-Agatoxin TK > 10μg