Learn More
Rotronic™ Water Activity Station Probe
Brand:  Rotronic™ HC2-AW
Product Code. 13558550
  • 2085.00€ / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
± 0.008 aw / 0.8 %rh / ±0.1 K
Stainless Steel, Aluminum
-40°C to +85°C, 0 to 100% RH