Learn More
Water, Advanced HPLC Grade for HPLC Gradient Analysis, Fisher Chemical™ Product Code.: 10221712
Water, Advanced HPLC Grade for HPLC Gradient Analysis, Fisher Chemical™
Product Code. 10221712 Shop All Fisher Chemical Products
2.5 L
Click to view available options
Quantity:
1 L
2.5 L
Product Code. 10221712
Brand: Fisher Chemical W/0110/PB17
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Request Bulk or Custom Format
For use with HPLC gradient analysis.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Plastic Coated Glass Bottle
Quantity 2.5 L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2539391 Certificate TypeCertificate of Analysis Date29/05/2025 Catalog NumberW/0110/PB15,W/0110/PB17
Lot Number2538745 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberW/0110/PB15,W/0110/PB17
Lot Number2538179 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberW/0110/PB15,W/0110/PB17
Lot Number2537624 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberW/0110/PB15,W/0110/PB17
Lot Number2536988 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberW/0110/PB15,W/0110/PB17
Product Identifier
  • Water
Product Content Correction
Product Title
Water, Advanced HPLC Grade for HPLC Gradient Analysis, Fisher Chemical™ >