Learn More
Water, HPLC for Fluorescence Detection, Fisher Chemical Product Code.: 10637042
Water, HPLC for Fluorescence Detection, Fisher Chemical
Product Code. 10637042 Shop All Fisher Chemical Products
2.5L, Amber glass bottle
Click to view available options
2.5L, Amber glass bottle
Product Code. 10637042
Brand: Fisher Chemical W/0107/17
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
HPLC for fluorescence detection.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Amber Glass Bottle
Quantity 2.5 L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2538745 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberW/0107/17
Lot Number2538179 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberW/0107/17
Lot Number2537624 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberW/0107/17
Lot Number2536988 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberW/0107/17
Lot Number2536798 Certificate TypeCertificate of Analysis Date24/05/2025 Catalog NumberW/0107/17
Product Identifier
  • Water
Product Content Correction
Product Title
Water, HPLC for Fluorescence Detection, Fisher Chemical >