Learn More
BINDER™ Water Supply
Water Supply for refillable tubular bags including peristaltic tubing, connectors, and fine gas filters.
Brand:  BINDER™ 8009-1083
 View more versions of this product
Product Code. 16464019
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
  • Water supply for versions of CO2 Incubators with active humidification
  • Sets consist of empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
Specifications
Specifications
Water supply - for versions with active humidification, Set consisting of 3 empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
Plastic
Set consisting of 3 empty, refillable tubular bags including peristaltic tubing, connectors, and fine gas filters
BINDER Chambers CBF Series CO2 Incubators with Hot Air Sterilization and Humidity Regulation
Water Supply