Learn More
ITS Thorsten Betzel™ WaterWorks™ Free Chlorine High Range
Use for determining whether or not you have the correct amount of sanitizer present. ITS Europe™ WaterWorks™ Free Chlorine High Range Strips combine a wide range of detection with patented Free Chlorine indicator.
Brand:  ITS Thorsten Betzel™ 480022
Product Code. 11722947
  • 18.90€ / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Wide range of detection
  • Free Chlorine indicator
  • Designed to resist interference from monochloramines
  • Perfect for testing in a food processing environment
  • Range of detection, from 0-120 ppm (mg/L)
  • Perfect for determining whether or not you have the correct amount of sanitizer present.
  • Perfect to take directly out onto the shop floor; thus reducing the amount of rejections and misadjusted processes.
Specifications
Specifications
Free Chlorine/1.0-5.0-10-20-40-80-120ppm
Bottle
50 Strips