Learn More
Thermo Scientific™ WebSeal™ 96-Well Mats for WebSeal Glass Inserted Plate Kits
Extend the usefulness of your well plate kits with replacement mats.
Brand:  Thermo Scientific™ 60180-M156
 View more versions of this product
Product Code. 15142789
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Extend the usefulness of your wellplate kits with WebSeal 96-Well Mats, which serve as accessories for WebSeal Glass Inserted Plate Kits.
Extend the usefulness of your wellplate kits with WebSeal 96-Well Mats, which serve as accessories for WebSeal Glass Inserted Plate Kits.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Welled Plugs, Pre-slit
96
WebSeal Glass Inserted Plate Kit (Cat. No. 60180-K108)
96-well WebSeal Mat
WebSeal Glass Inserted 96-well Plate
Silicone, PTFE
1 Mat
Yes
Webseal