Learn More
Thermo Scientific™ WebSeal™ Sealing Tapes
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes.
Brand: Thermo Scientific™ 60180-M143
197.89 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes. Available in a selection of thin, lightweight materials and adhesives to meet the needs of any application or assay, they minimize evaporation and protect samples from contamination and spilling.
Easy, Consistent Sealing
- Pressure-sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Tapes seal onto the plate with a convenient, handheld applicator (available separately)
Specifications
Clear | |
100 | |
Webseal | |
-70°C to +100°C |
PES, Silicone | |
Non-sterile Sealing Tape | |
Pressure Sensitive | |
All Microplates |
Produktvorschläge
Customers who viewed this item also viewed
Sonderangebote
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
5Hours
21Minutes
18Seconds
17