Learn More
Thermo Scientific™ WebSeal™ Sealing Tapes
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes.
Brand: Thermo Scientific™ 60180-M145
188.26 EUR valid until 2025-12-31
Use promo code "24292" to get your promotional price.
Description
Effectively seal all microplate formats with Thermo Scientific™ WebSeal™ Sealing Tapes. Available in a selection of thin, lightweight materials and adhesives to meet the needs of any application or assay, they minimize evaporation and protect samples from contamination and spilling.
Easy, Consistent Sealing
- Pressure-sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
- Tapes seal onto the plate with a convenient, handheld applicator (available separately)
Specifications
Clear | |
Polyolefin, Rubber | |
Non-sterile Sealing Tape | |
Pressure Sensitive |
Sealing Tape with SRA (synthetic rubber adhesive) | |
100 | |
Webseal | |
All Microplates |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
5Hours
21Minutes
18Seconds
10