Learn More
Thermo Scientific™ Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments
Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments are for use with ACC- and WPS-3000 autosamplers.
907.00€
Specifications
Type | Low Profile Support Rack |
---|---|
For Use With (Equipment) | Deep-Well Plates for All WPS-3000 LC Autosamplers (Not ACC) |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13472018
|
Thermo Scientific™
6820.4079 |
Support rack for deep-well plate, 34-46 mm height for all WPS-3000 variants |
907.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Use Thermo Scientific™ Dionex™ Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments with ACC- and WPS-3000 autosamplers.
Specifications
Low Profile Support Rack | |
Deep-Well Plates for All WPS-3000 LC Autosamplers (Not ACC) |
Product Title
Thermo Scientific™ Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments