Learn More
Cytiva Whatman™ Membrane Holder, glass reservoir
Vacuum-Type Glass Membrane Holder, glass reservoir, 47 mm, 300 ml volume, membrane filter accessories
Brand: Cytiva 1961-054
161.50 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Borosilicate Glass | |
Vacuum-Type Glass Membrane Holder | |
300 mL | |
47 mm |
Product Content Correction
Product Title
Cytiva Whatman™ Membrane Holder, glass reservoir > Glass reservoir holder