Learn More
Cytiva Whatman™ pH Indicators Reel Dispensers
Click to view available options
pH Range:
0.5 to 5.5
1 to 11
1 to 14
4 to 7
6.4 to 8
8 to 10
Unit Size:
Each
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
Specifications
| pH Range | 4 to 7 |
| Format | Reel |
| For Use With (Application) | Routine pH testing |
| Type | pH Indicator Dispenser |