Learn More
DWK Life Sciences Wheaton™ 25mm Filtration Assemblies and Accessories
Choose from a wide range of filtration assemblies and accessories to suit your application needs
71.05€ - 248.00€
Specifications
For Use With (Equipment) 25mm filtration assembly
View More Specs
Products 5
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11754309
View Documents
DWK Life Sciences
419336
Silicone Stopper No. 5
99.80€
Pack of 6
In Stock
to see stock.
Add to basket
12451571
View Documents
DWK Life Sciences
419325
Filtration Assembly
248.00€
Each
In Stock
to see stock.
Add to basket
12461571
View Documents
DWK Life Sciences
419330
Funnel
71.05€
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
12481571
View Documents
DWK Life Sciences
419332
Fritted Glass Base
129.00€
Each
Estimated Shipment: 09-09-2025
to see stock.
Add to basket
12487121
View Documents
DWK Life Sciences
635229
Filtration Flask
73.90€
Each
Estimated Shipment: 09-09-2025
Specifications
25mm filtration assembly
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ 25mm Filtration Assemblies and Accessories