Learn More
DWK Life Sciences WHEATON™ Dropper Assembly for 7 mL Clear Type 1B Tubular Glass Dropper Bottle
Use promo code "23259" to get your promotional price.
Description
WHEATON™ glass dropper bottles and assemblies are manufactured from clear neutral tubular glass and are the perfect primary packaging solution for a range of diagnostic, pharmaceutical and healthcare applications. Made from chemically inert Type 1B Neutral Glass, the glass dropper bottles and dropper assemblies are suitable for key packaging applications such as blood grouping reagents and rare grouping sera. Glass dropper bottles are also an advantage for several in vitro diagnostic applications where false positives from plastic dropper bottles may be a concern. The dropper assemblies are manufactured with pharmaceutical grade rubber bulbs and a choice of coloured caps to help differentiate products in kits, and are also manufactured with a controlled drop size for consistency. The vials are also available to purchase either as trays of vials packed in a box, or trays of vials shrink wrapped onto a pallet to reduce regular shipping costs.
- Manufactured from Type 1B Clear Neutral Glass
- Controlled drop size dropper assembly
- Choice of different coloured caps on dropper assemblies allows for differentiation in diagnostic kits
Spécification
Description | Black Bulb |
Sterility | Non-sterile |
Closure Color | Red |
Autoclavable | Autoclavable |
Disposable | Single-use |
Drop Size | 45 to 55 μL |
Closure Material | Polypropylene |