Learn More
DWK Life Sciences Wheaton™ Glass Slide Rack
Made of sode-lime glass.
Brand: DWK Life Sciences 900304
236.28 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
Glass slide rack:
- Holds 20-40 slides
- 20 single slides
- 39 slides arranged alternately straight across and diagonally
- 40 slides back-to-back
- Accommodates standard slides
- 3 x 1 in. (75 x 25mm)
- 3 x 1.5 in. (75 x 38mm)
- 3 x 2 in. (75 x 51mm)
- Hinged handle
- Folds neatly inside dish to allow cover to be placed on top
- Enables easy insertion and removal of slides
Specifications
Soda Lime Glass | |
Standard Microscope Slides, Staining Dish | |
20 to 40 | |
4.6 in. | |
3.4 in. | |
2.7 in. |
Slide Rack | |
Hinged, Stainless Steel | |
4.6 x 3.4 in. (11.8 x 8.8 cm) | |
11.8 cm | |
8.8 cm | |
7 cm |
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Glass Slide Rack > Slide rack with handle; For 20-40 slides