missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Media/Laboratory Bottle Caps

Product Code. 12664803 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
33-430
38-430
Material:
Polyethylene
Rubber
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with Wheaton media bottles

  • Black phenolic caps with screw thread
  • Low Density Polyethylene (Not autoclavable), Rubber lined (Autoclavable)

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Polyethylene
No. per Case 200 Cs.
Neck Finish 38-430
Product Type Media Bottle Cap
For Use With (Equipment) Media bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.