Learn More
DWK Life Sciences Wheaton™ Round Translucent LDPE Bottles
Click to view available options
Capacity:
0.24 oz., 7 mL
0.5 oz., 15 mL
1 oz., 30mL
Height (English):
2.4 in.
2.87 in.
3.07 in.
Height (Metric):
63 mm
73 mm
78 mm
Neck Finish:
15-415
20-410
Description
Narrow mouth is ideal for liquids.
- Excellent for mild and strong buffers, good chemical resistance
- Pre-attached white polypropylene screw cap with foamed polyethylene liner
Specifications
Specifications
Capacity | 1 oz., 30mL |
Capacity (Metric) | 30 mL |
Capacity (English) | 1 oz. |
Color | Translucent |
Diameter (English) | 1.29 in. |
Material | LDPE |
Closure Color | White |
Neck Finish | 20-410 |
Certifications/Compliance | ISO 9001 |
Closure Included | Yes |